![Fmoc-Pro-OPfp Novabiochem®](/deepweb/assets/sigmaaldrich/product/images/709/572/250b668a-354a-4b0d-b571-5d7596065b79/640/250b668a-354a-4b0d-b571-5d7596065b79.jpg)
Fmoc-Pro-OPfp
![Fmoc-Pro-OPfp Novabiochem®](/deepweb/assets/sigmaaldrich/product/images/709/572/250b668a-354a-4b0d-b571-5d7596065b79/640/250b668a-354a-4b0d-b571-5d7596065b79.jpg)
Synonym(s):
Fmoc-Pro-OPfp
Empirical Formula (Hill Notation):
C26H18F5NO4
CAS Number:
Molecular Weight:
503.42
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Functional Group
Application
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing