Skip to Content
MilliporeSigma
Fmoc-Glu(OtBu)-OH ≥98.0% (HPLC)

Fmoc-Glu(OtBu)-OH

Fmoc-Glu(OtBu)-OH ≥98.0% (HPLC)
Synonym(s):
Fmoc-L-glutamic acid 5-tert-butyl ester, Fmoc-Glu(OtBu)-OH
Empirical Formula (Hill Notation):
C24H27NO6
CAS Number:
Molecular Weight:
425.47

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing