Flurbiprofen for peak identification
Synonym(s):
Flurbiprofen, (±)-2-Fluoro-α-methyl-4-biphenylacetic acid, L-790,330
Linear Formula:
C6H5C6H3(F)CH(CH3)CO2H
CAS Number:
Molecular Weight:
244.26
Products
Formula Weight
Manufacturer
Melting Point (°C)
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing