Skip to Content
MilliporeSigma
Fluorescamine ≥98% (TLC), powder, used for detection of primary amines

Fluorescamine

Fluorescamine ≥98% (TLC), powder, used for detection of primary amines
Synonym(s):
4-Phenylspiro-[furan-2(3H),1-phthalan]-3,3′-dione
Empirical Formula (Hill Notation):
C17H10O4
CAS Number:
Molecular Weight:
278.26
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing