Skip to Content
MilliporeSigma
vibrant-m

Fluocortolone pivalate for ID and assay

vibrant-m
Synonym(s):
Fluocortolone pivalate, (6α,11β,16α)-21-(2,2-Dimethyl-1-oxopropoxy)-6-fluoro-11-hydroxy-16-methylpregna-1,4-diene-3,20-dione
Empirical Formula (Hill Notation):
C27H37FO5
CAS Number:
Molecular Weight:
460.58

Products

Formula Weight
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1