Skip to Content
MilliporeSigma
Flunixin meglumine European Pharmacopoeia (EP) Reference Standard

Flunixin meglumine

Flunixin meglumine European Pharmacopoeia (EP) Reference Standard
Synonym(s):
2-[[2-Methyl-3-(trifluoromethyl)phenyl]amino]-3-pyridinecarboxylic acid meglumine salt, Banamine, Flunixin meglumine
Empirical Formula (Hill Notation):
C14H11F3N2O2·C7H17NO5
CAS Number:
Molecular Weight:
491.46

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique