Flumetralin
Empirical Formula (Hill Notation):
C16H12ClF4N3O4
CAS Number:
Molecular Weight:
421.73
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2