Skip to Content
MilliporeSigma
Fluconazole solution 2.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Fluconazole solution

Fluconazole solution 2.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C13H12F2N6O
CAS Number:
Molecular Weight:
306.27
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Format
Application
Package Size
Reference Material Type
Special Grade
Technique