Skip to Content
MilliporeSigma
Flucloxacillin sodium VETRANAL®, analytical standard

Flucloxacillin sodium

Flucloxacillin sodium VETRANAL®, analytical standard
Synonym(s):
(2S,5R,6R)-6-[[[3-(2-Chloro-6-fluorophenyl)-5-methyl-4-isoxazolyl]carbonyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid sodium, BRL 2039 sodium, FK 900 sodium, Floxacillin sodium
Empirical Formula (Hill Notation):
C19H16ClFN3NaO5S
CAS Number:
Molecular Weight:
475.85
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique