Skip to Content
MilliporeSigma
vibrant-m

Fenticonazole Nitrate

vibrant-m
Synonym(s):
Fenticonazole nitrate
Empirical Formula (Hill Notation):
C24H21Cl2N3O4S
CAS Number:
Molecular Weight:
518.41

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing