Skip to Content
MilliporeSigma
Fenoxaprop-ethyl PESTANAL®, analytical standard

Fenoxaprop-ethyl

Fenoxaprop-ethyl PESTANAL®, analytical standard
Synonym(s):
Ethyl 2-{4-[(6-chlorobenzoxazol-2-yl)oxy]phenoxy}propionate
Empirical Formula (Hill Notation):
C18H16ClNO5
CAS Number:
Molecular Weight:
361.78
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing