Skip to Content
MilliporeSigma
Fast Green FCF analytical standard

Fast Green FCF

Fast Green FCF analytical standard
Synonym(s):
Food green 3
Linear Formula:
C37H34N2O10S3Na2
CAS Number:
Molecular Weight:
808.85
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Chemical Composition
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3