Skip to Content
MilliporeSigma
exo-THC solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

exo-THC solution

exo-THC solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Synonym(s):
exo-THC solution
Empirical Formula (Hill Notation):
C21H30O2
CAS Number:
Molecular Weight:
314.46
EC Number:

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1