Skip to Content
MilliporeSigma
Exo-4-anisole Kwon [2.2.1] bicyclic phosphine

Exo-4-anisole Kwon [2.2.1] bicyclic phosphine

Exo-4-anisole Kwon [2.2.1] bicyclic phosphine
Synonym(s):
Exo-4-Methoxyphenyl Kwon [2.2.1] Bicyclic Phosphine, (1S,4S,5R)-5-(4-methoxyphenyl)-2-tosyl-2-aza-5-phosphabicyclo[2.2.1]heptane
Empirical Formula (Hill Notation):
C19H22NO3PS
Molecular Weight:
375.42

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing