![EX-CELL® CD CHO Serum-Free Medium for CHO Cells, Chemically Defined Animal-component free, without L-glutamine, without hypoxanthine, without thymidine, liquid, sterile-filtered, suitable for cell culture](/deepweb/assets/sigmaaldrich/product/images/410/339/a0ed2104-1d4b-4854-8ca5-2ab50d374125/640/a0ed2104-1d4b-4854-8ca5-2ab50d374125.jpg)
EX-CELL® CD CHO Serum-Free Medium for CHO Cells, Chemically Defined
![EX-CELL® CD CHO Serum-Free Medium for CHO Cells, Chemically Defined Animal-component free, without L-glutamine, without hypoxanthine, without thymidine, liquid, sterile-filtered, suitable for cell culture](/deepweb/assets/sigmaaldrich/product/images/410/339/a0ed2104-1d4b-4854-8ca5-2ab50d374125/640/a0ed2104-1d4b-4854-8ca5-2ab50d374125.jpg)
Synonym(s):
EX-CELL® CD CHO Serum-Free Medium for CHO Cells, Chemically Defined
Products
Product Category
Brand
Physical Form
Product Line
HEPES (mM)
L-Glutamine
Phenol Red
Quality Segment
Sterilization Method
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing