
Eukitt® Quick-hardening mounting medium

Synonym(s):
Poly(butyl methacrylate-co-methyl methacrylate)
Linear Formula:
[CH2C(CH3)[CO2(CH2)3CH3]]X[CH2C(CH3)(CO2CH3)]Y
CAS Number:
Products
Color
Physical Form
Quality Segment
Application
Showing 1-1 of 1