![Eucalyptol phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/341/464/c2116f21-43f3-45c3-a888-36026d5d95f0/640/c2116f21-43f3-45c3-a888-36026d5d95f0.png)
Eucalyptol
![Eucalyptol phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/341/464/c2116f21-43f3-45c3-a888-36026d5d95f0/640/c2116f21-43f3-45c3-a888-36026d5d95f0.png)
Synonym(s):
1,3,3-Trimethyl-2-oxabicyclo[2.2.2]octane, 1,8-Cineole, 1,8-Epoxy-p-menthane
Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Products
Product Category
Brand
Boiling Point (°C)
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5