![Ethynyl p-tolyl sulfone 98%](/deepweb/assets/sigmaaldrich/product/structures/451/506/86e2b613-ea61-4cc2-a58d-18f9ae291cab/640/86e2b613-ea61-4cc2-a58d-18f9ae291cab.png)
Ethynyl p-tolyl sulfone
![Ethynyl p-tolyl sulfone 98%](/deepweb/assets/sigmaaldrich/product/structures/451/506/86e2b613-ea61-4cc2-a58d-18f9ae291cab/640/86e2b613-ea61-4cc2-a58d-18f9ae291cab.png)
Synonym(s):
p-Toluenesulfonylacetylene, Tosylacetylene
Linear Formula:
CH3C6H4SO2C≡CH
CAS Number:
Molecular Weight:
180.22
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing