Skip to Content
MilliporeSigma
Ethyl vinyl ether contains 0.1% KOH as stabilizer, 99%

Ethyl vinyl ether

Ethyl vinyl ether contains 0.1% KOH as stabilizer, 99%
Synonym(s):
Ethoxyethylene, Ethyl vinyl ether
Linear Formula:
C2H5OCH=CH2
CAS Number:
Molecular Weight:
72.11

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing