Skip to Content
MilliporeSigma
Ethyl vanillin Pharmaceutical Secondary Standard; Certified Reference Material

Ethyl vanillin

Ethyl vanillin Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
3-Ethoxy-4-hydroxybenzaldehyde, Ethylvanillin, NSC 1803, NSC 67240
Linear Formula:
C2H5OC6H3(OH)CHO
CAS Number:
Molecular Weight:
166.17
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique