Skip to Content
MilliporeSigma
Ethyl-tetramethylcyclopentadiene 97%

Ethyl-tetramethylcyclopentadiene

Ethyl-tetramethylcyclopentadiene 97%
Empirical Formula (Hill Notation):
C11H18
CAS Number:
Molecular Weight:
150.26

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing