Skip to Content
MilliporeSigma
Ethyl (S)-(−)-4-chloro-3-hydroxybutyrate 96%

Ethyl (S)-(−)-4-chloro-3-hydroxybutyrate

Ethyl (S)-(−)-4-chloro-3-hydroxybutyrate 96%
Linear Formula:
ClCH2CH(OH)CH2CO2C2H5
CAS Number:
Molecular Weight:
166.60

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing