Skip to Content
MilliporeSigma
Ethyl (methylthio)acetate ≥98%, FG

Ethyl (methylthio)acetate

Ethyl (methylthio)acetate ≥98%, FG
Synonym(s):
(Methylthio)acetic acid ethyl ester, Ethyl α-(methylthio)acetate
Linear Formula:
CH3SCH2CO2C2H5
CAS Number:
Molecular Weight:
134.20
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Color
Formula Weight
Physical Form
Purity
Agency
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing