Skip to Content
MilliporeSigma
Ethylene di(p-toluenesulfonate) 97%

Ethylene di(p-toluenesulfonate)

Ethylene di(p-toluenesulfonate) 97%
Synonym(s):
Ethylene ditosylate, Ethylene glycol ditosylate
Linear Formula:
[CH3C6H4SO3CH2-]2
CAS Number:
Molecular Weight:
370.44

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing