Skip to Content
MilliporeSigma
Ethylenediamine diacetate 98%

Ethylenediamine diacetate

Ethylenediamine diacetate 98%
Linear Formula:
NH2CH2CH2NH2 · 2CH3COOH
CAS Number:
Molecular Weight:
180.20

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing