Skip to Content
MilliporeSigma
Ethylenediamine meets USP testing specifications

Ethylenediamine

Ethylenediamine meets USP testing specifications
Synonym(s):
1,2-Diaminoethane
Linear Formula:
NH2CH2CH2NH2
CAS Number:
Molecular Weight:
60.10
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Feature
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Reagent Type
Reference Material Type
Special Grade
Technique