Skip to Content
MilliporeSigma
Ethyl benzoate analytical standard

Ethyl benzoate

Ethyl benzoate analytical standard
Linear Formula:
C6H5COOC2H5
CAS Number:
Molecular Weight:
150.17
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing