Skip to Content
MilliporeSigma
Ethyl arachidonate ≥98.5% (GC), liquid

Ethyl arachidonate

Ethyl arachidonate ≥98.5% (GC), liquid
Synonym(s):
5,8,11,14-Eicosatetraenoic acid ethyl ester, Arachidonic acid ethyl ester
Empirical Formula (Hill Notation):
C22H36O2
CAS Number:
Molecular Weight:
332.52

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing