Skip to Content
MilliporeSigma
Ethyl 4-(dimethylamino)benzoate suitable for qNMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Ethyl 4-(dimethylamino)benzoate

Ethyl 4-(dimethylamino)benzoate suitable for qNMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Ethyl 4-dimethylaminobenzoate
Linear Formula:
(CH3)2NC6H4CO2C2H5
CAS Number:
Molecular Weight:
193.24
EC Number:

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique