Skip to Content
MilliporeSigma
Ethyl 3-methyl-2-oxobutyrate 97%

Ethyl 3-methyl-2-oxobutyrate

Ethyl 3-methyl-2-oxobutyrate 97%
Synonym(s):
Ethyl dimethylpyruvate, Ketovaline ethyl ester
Linear Formula:
(CH3)2CHCOCOOCH2CH3
CAS Number:
Molecular Weight:
144.17
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing