Skip to Content
MilliporeSigma
Ethyl 3-hydroxybutyrate ≥98%

Ethyl 3-hydroxybutyrate

Ethyl 3-hydroxybutyrate ≥98%
Linear Formula:
CH3CH(OH)CH2COOC2H5
CAS Number:
Molecular Weight:
132.16
EC Number:

Products

Product Category
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing