Skip to Content
MilliporeSigma
Ethyl (2S)2-[(3S,5aS,8aS, 9aS)-3-methyl-1,4-dioxodecahydro-1H-cyclopenta[e]pyrrolo[1,2-a]pyrazin-2-yl]-4-phenylbutanoate certified reference material, pharmaceutical secondary standard

Ethyl (2S)2-[(3S,5aS,8aS, 9aS)-3-methyl-1,4-dioxodecahydro-1H-cyclopenta[e]pyrrolo[1,2-a]pyrazin-2-yl]-4-phenylbutanoate

Ethyl (2S)2-[(3S,5aS,8aS, 9aS)-3-methyl-1,4-dioxodecahydro-1H-cyclopenta[e]pyrrolo[1,2-a]pyrazin-2-yl]-4-phenylbutanoate certified reference material, pharmaceutical secondary standard
Synonym(s):
Ramipril diketopiperazine
Empirical Formula (Hill Notation):
C23H30N2O4
CAS Number:
Molecular Weight:
398.50

Products

Brand
Formula Weight
Quality Segment
Application
Package Size
API Family
Special Grade