Skip to Content
MilliporeSigma
Ethyl 2-methylbutyrate ≥98%, FCC, FG

Ethyl 2-methylbutyrate

Ethyl 2-methylbutyrate ≥98%, FCC, FG
Linear Formula:
CH3CH2CH(CH3)COOC2H5
CAS Number:
Molecular Weight:
130.18
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3