Skip to Content
MilliporeSigma
Ethyl 1H-imidazole-1-carboxylate

Ethyl 1H-imidazole-1-carboxylate

Ethyl 1H-imidazole-1-carboxylate
Synonym(s):
N-Carboethoxyimidazole, EtImC, Ethyl imidazolecarbamate, Sarpong reagent
Empirical Formula (Hill Notation):
C6H8N2O2
CAS Number:
Molecular Weight:
140.14
EC Number:

Products

Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing