Skip to Content
MilliporeSigma
Ethyl 1-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride technical grade

Ethyl 1-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride

Ethyl 1-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride technical grade
Synonym(s):
1-Benzyl-4-ethoxycarbonyl-3-piperidone hydrochloride
Empirical Formula (Hill Notation):
C15H19NO3 · HCl
CAS Number:
Molecular Weight:
297.78
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing