Skip to Content
MilliporeSigma
Ethephon certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Ethephon

Ethephon certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
2-Chloroethylphosphonic acid, Ethephon
Linear Formula:
ClCH2CH2P(O)(OH)2
CAS Number:
Molecular Weight:
144.49

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique