Skip to Content
MilliporeSigma
Ethanesulfonic acid 95%

Ethanesulfonic acid

Ethanesulfonic acid 95%
Synonym(s):
Ethanesulfonic acid
Linear Formula:
CH3CH2SO3H
CAS Number:
Molecular Weight:
110.13

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing