Skip to Content
MilliporeSigma
Estriol 16α-(β-D-glucuronide) ≥97%

Estriol 16α-(β-D-glucuronide)

Estriol 16α-(β-D-glucuronide) ≥97%
Synonym(s):
1,3,5(10)-Estratriene-3,16α,17β-triol 16-glucuronide, 3,16α,17β-Trihydroxy-1,3,5(10)-estratriene 16-glucuronide
Empirical Formula (Hill Notation):
C24H32O9
CAS Number:
Molecular Weight:
464.51

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Research Area
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing