Skip to Content
MilliporeSigma
Erythromycin tested according to Ph. Eur.

Erythromycin

Erythromycin tested according to Ph. Eur.
Empirical Formula (Hill Notation):
C37H67NO13
CAS Number:
Molecular Weight:
733.93
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Technique