Skip to Content
MilliporeSigma
Ergotamine tartrate European Pharmacopoeia (EP) Reference Standard

Ergotamine tartrate

Ergotamine tartrate European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Ergotamine D-tartrate
Empirical Formula (Hill Notation):
C33H35N5O5 · 0.5C4H6O6
CAS Number:
Molecular Weight:
656.70

Products

Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade