![Ergocalciferol for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/319/554/6d1e5e74-2616-4acb-96f2-47249a5007ac/640/6d1e5e74-2616-4acb-96f2-47249a5007ac.png)
Ergocalciferol for system suitability
![Ergocalciferol for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/319/554/6d1e5e74-2616-4acb-96f2-47249a5007ac/640/6d1e5e74-2616-4acb-96f2-47249a5007ac.png)
Synonym(s):
Ergocalciferol, Calciferol, Ercalciol, Ergosterol irradiated, Irradiated ergosterol, Vitamin D2
Empirical Formula (Hill Notation):
C28H44O
CAS Number:
Molecular Weight:
396.65
Products
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing