Skip to Content
MilliporeSigma
Epidermal Growth Factor, human, animal component free EGF, recombinant, expressed in Escherichia coli, >97% (SDS-PAGE)

Epidermal Growth Factor, human, animal component free

Epidermal Growth Factor, human, animal component free EGF, recombinant, expressed in Escherichia coli, >97% (SDS-PAGE)
Synonym(s):
Epidermal Growth Factor human, EGF
CAS Number:

Products

Product Category
Brand
Biological Source
Color
Gene Alias
Physical Form
Purity
Recombinant Host
Quality Segment