Skip to Content
MilliporeSigma
Eosin Y disodium salt certified by the Biological Stain Commission

Eosin Y disodium salt

Eosin Y disodium salt certified by the Biological Stain Commission
Synonym(s):
2′,4′,5′,7′-Tetrabromofluorescein disodium salt, Acid Red 87, Bromo acid J. TS, XL, or XX, Bromofluorescein, Bronze Bromo ES, D&C;Red No. 22, Eosin yellowish
Empirical Formula (Hill Notation):
C20H6Br4Na2O5
CAS Number:
Molecular Weight:
691.85
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application