Skip to Content
MilliporeSigma
Enalapril maleate salt powder, ≥98% (TLC)

Enalapril maleate salt

Enalapril maleate salt powder, ≥98% (TLC)
Synonym(s):
(S)-N-[1-(Ethoxycarbonyl)-3-phenylpropyl]-Ala-Pro maleate salt
Empirical Formula (Hill Notation):
C20H28N2O5 · C4H4O4
CAS Number:
Molecular Weight:
492.52
EC Number:

Products

Product Category
Color
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing