Skip to Content
MilliporeSigma
EMPIGEN® BB detergent ~30% active substance

EMPIGEN® BB detergent

EMPIGEN® BB detergent ~30% active substance
Synonym(s):
N,N-Dimethyl-N-dodecylglycine betaine, N-(Alkyl C10-C16)-N,N-dimethylglycine betaine
Linear Formula:
CH3(CH2)8-14CH2N+(CH3)2CH2COO-
CAS Number:
EC Number:

Products

Product Category
Brand
Physical Form
Chemical Composition
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing