Skip to Content
MilliporeSigma
Emodin 8-glucoside phyproof® Reference Substance

Emodin 8-glucoside

Emodin 8-glucoside phyproof® Reference Substance
Synonym(s):
1,6-dihydroxy-3-methyl-8-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyanthracene-9,10-dione, Anthraglycoside B, Emodin 8-β-D-Glucopyranoside, Emodin glucoside B
Empirical Formula (Hill Notation):
C21H20O10
CAS Number:
Molecular Weight:
432.38

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing