Skip to Content
MilliporeSigma
EHNA hydrochloride ≥98% (HPLC)

EHNA hydrochloride

EHNA hydrochloride ≥98% (HPLC)
Synonym(s):
erythro-9-(2-Hydroxy-3-nonyl)­adenine hydrochloride, erythro-9-Amino-β-hexyl-­α-methyl-9H-purine-9-ethanol hydrochloride
Empirical Formula (Hill Notation):
C14H23N5O · HCl
CAS Number:
Molecular Weight:
313.83

Products

Product Category
Formula Weight
Gene Alias
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing