![ECO Brij® S100 average Mn ~4,670](/deepweb/assets/sigmaaldrich/product/structures/214/322/d296dd36-df8d-4eb1-b11f-4cc92ab5d6cb/640/d296dd36-df8d-4eb1-b11f-4cc92ab5d6cb.png)
ECO Brij® S100
![ECO Brij® S100 average Mn ~4,670](/deepweb/assets/sigmaaldrich/product/structures/214/322/d296dd36-df8d-4eb1-b11f-4cc92ab5d6cb/640/d296dd36-df8d-4eb1-b11f-4cc92ab5d6cb.png)
Synonym(s):
Brij® S 100, Polyoxyethylene (100) stearyl ether
Linear Formula:
C18H37(OCH2CH2)nOH, n~100
CAS Number:
Products
Brand
Sustainability
Feature
Melting Point (°C)
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing