Skip to Content
MilliporeSigma
[D-Trp7, Ala8, D-Phe10]-α-Melanocyte Stimulating Hormone Amide Fragment 6-11 ≥97% (HPLC)

[D-Trp7, Ala8, D-Phe10]-α-Melanocyte Stimulating Hormone Amide Fragment 6-11

[D-Trp7, Ala8, D-Phe10]-α-Melanocyte Stimulating Hormone Amide Fragment 6-11 ≥97% (HPLC)
Synonym(s):
GHRP-6
Empirical Formula (Hill Notation):
C46H56N12O6
CAS Number:
Molecular Weight:
873.01

Products

Product Category
Brand
Formula Weight
Gene Alias
Purity
Quality Segment
Showing 1-1 of 1