Drospirenone impurity E
Synonym(s):
17-epi-Drospirenone
Empirical Formula (Hill Notation):
C24H30O3
CAS Number:
Molecular Weight:
366.49
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing